Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrabutylammonium sulfate 50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Warning

41723-K4 41723-K4 41723-AE 41723-AE 41723-AP 41723-AP
AA41723-AEEA 51.69 USD InStock
AA41723-AE CAAA41723-AE AA41723-AP CAAA41723-AP AA41723-K4 CAAA41723-K4
Tetrabutylammonium sulfate 50% (w/w) in aqueous solution
Tetrabutylammonium sulphate
Applications: Phase transfer catalyst, neutral pH

Phase transfer catalyst, neutral pH


Formula: C₃₂H₇₂N₂O₄S
Boiling Pt: 84 °C
Density: 1,01
Storage Temperature: Ambient
MDL Number: MFCD00216632
CAS Number: 2472-88-0
EINECS: 219-593-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty