Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Humic acid ≥98%

Supplier: Thermo Scientific Chemicals

041747-06 41747-14 41747-06 041747-14
AA41747-06EA 69.74 USD InStock
CAAA41747-06 AA41747-06 CAAA41747-14 AA41747-14
Humic acid ≥98%
Humic acid
Applications: Metal chelator; pigments
Slightly soluble in water, with much swelling


Formula: C9H9NO6
Storage Temperature: Ambient
MDL Number: MFCD00147177
CAS Number: 1415-93-6
EINECS: 215-809-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty