Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tellurite(IV) ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Sodium telluriteDisodium trioxotellurate

41778-18 041778-09 41778-09 041778-18
AA41778-18EA 232.99 USD InStock
CAAA41778-09 CAAA41778-18 AA41778-18 AA41778-09
Sodium tellurite(IV) ≥99.5% (metals basis)
Sodium tellurite(IV)


Formula: Na₂O₃Te
MW: 221.58 g/mol
Melting Pt: 710 °C
Storage Temperature: Ambient
MDL Number: MFCD00036141
CAS Number: 10102-20-2
EINECS: 233-268-4
UN: 3284
ADR: 6.1,II
Merck Index: 12,08834
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty