Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Molybdenum(IV) sulfide ≥98% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Molybdenum disulphide

41827-22 041827-22 41827-36 41827-14 041827-14 041827-36
AA41827-22EA 104.52 USD InStock
AA41827-22 CAAA41827-36 CAAA41827-14 CAAA41827-22 AA41827-14 AA41827-36
Molybdenum(IV) sulfide ≥98% (metals basis)
Molybdenum (IV) sulphide
Molybdenum(IV) sulfide is used as a catalyst in the electrolysis of water. It is employed as a cocatalyst for desulfurization in petrochemistry especially hydrodesulfurization. It is also used as a component of photoelectrochemical and microelectronics applications. Further, it is useful as a filament material in many electrical applications. It finds application as a lubricant in aircraft engines. Polymers consisting of molybdenum(IV) sulfide such as nylon, teflon and vespel posses improved strength and reduced friction.

  • -325 Mesh Powder


Formula: MoS₂
MW: 160.07 g/mol
Melting Pt: 1185 °C
Density: 4.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003470
CAS Number: 1317-33-5
EINECS: 215-263-9
Merck Index: 12,06318
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty