Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dicerium trisulfide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Cerium (III) sulphide Cerous sulphide

41856-14 41856-06 041856-14 041856-06
AA41856-06EA 43.69 USD InStock
AA41856-06 CAAA41856-06 AA41856-14 CAAA41856-14
Dicerium trisulfide ≥99.9% (metals basis)
Dicerium trisulphide


Formula: Ce₂S₃
MW: 376.43 g/mol
Melting Pt: 2100 °C
Density: 5,02
Storage Temperature: Ambient
MDL Number: MFCD00016007
CAS Number: 12014-93-6
EINECS: 234-603-7
UN: 3134
ADR: 4.3,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty