Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium citrate hydrate (∼ 16% Mg) (dried basis) (dry weight)

Supplier: Thermo Scientific Chemicals
Synonyms: Magnesium citrate tribasic hydrate

41921-A1 41921-B5 41921-A7 041921-A1 041921-A7 41921-B5
AA41921-A1EA 58.49 USD InStock
AA41921-A1 AAA41921-B5 AA41921-A7 CAAA41921-A7 CAAA41921-A1 CAAAA41921-B5
Magnesium citrate hydrate (∼ 16% Mg) (dried basis) (dry weight)
Magnesium citrate hydrate


Formula: C₁₂H₁₀Mg₃O₁₄·H₂O
MW: 451.13 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00036422
CAS Number: 120156-45-8
EINECS: 222-093-9
Order Now
Specification Test Results
Form Powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty