Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Magnesium citrate dibasic hydrate

Supplier: Thermo Scientific Chemicals

41954-A1 041954-A1 041954-30 41954-30 41954-A7
AA41954-30EA 107.53 USD InStock
AA41954-30 CAAA41954-30 AA41954-A1 CAAA41954-A1 CAAA41954-A7
Magnesium citrate dibasic hydrate
Magnesium citrate dibasic hydrate


Formula: C₆H₆MgO₇•xH₂O
MW: 214.412 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00151706
CAS Number: 119851-23-9
EINECS: 231-923-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty