Would you like to log in again?
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Supplier: Thermo Scientific Chemicals
Danger
42012-VA 042012-VA 042012-CF 42012-CF
AA42012-CFEA 268.99 USD InStock
AA42012-CF CAAA42012-CF AA42012-VA CAAA42012-VA
Stainless steel (Fe:Cr:Ni; 70:19:11 wt%), gauze woven from wire, twilled weave 400 mesh woven from 0.028mm (0.0011in) Type 304
Stainless steel
Open area: 31.4%; Width of opening: 0.0014in
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty