Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrahydrofuran, anhydrous ≥99.8% unstabilized

Supplier: Thermo Scientific Chemicals

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Danger
Synonyms: 1,4-epoxy-ButaneButane alpha,delta-oxideCyclotetramethylene oxideOxacyclopentaneOxolaneTetramethylene oxideTHF

042254-K2 042254-K7 042254-AK
AA42254-K2EA 146.24 USD InStock
AA42254-K2 AA42254-AK AA42254-K7
Tetrahydrofuran, anhydrous ≥99.8% unstabilized
Tetrahydrofuran
Applications: Synthesis and Liquid Chromotography


Formula: C₄H₈O
MW: 72.11 g/mol
Boiling Pt: 66 °C
Melting Pt: –108 °C
Density: 0.889
Flash Pt: –17 °C (1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/19/789519.jpg
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty