Would you like to log in again?
Fused Quartz Microscope Slides, Thermo Scientific
Supplier: Thermo Scientific
042297.KJ 042297.KG 042296.KG 42295-KJ 042296.KJ 042295.KG
AA42296-KGEA 49.43 USD InStock
AA42296-KG AA42295-KG AA42296-KJ AA42297-KJ AA42295-KJ AA42297-KG
Fused Quartz Microscope Slides, Thermo Scientific
Slides Microscope Slides
These quartz discs are manufactured by fusing naturally occuring crystalline silica.
  • High chemical and corrosion resistance
  • Ground and polished
  • High melting point
  • Extreme hardness
  • Low coeffiecent of thermal expansion
  • Excellent electrical insulation quality
  • Optical transmission from ultra-violet to infra-red
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty