Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold plating solution ∼3.7 g/l metal content, electroless

Supplier: Thermo Scientific Chemicals
Danger

042307.K2 042307.AK 42307-K2 42307-AK
AA42307-AKEA 275.85 USD InStock
AA42307-AK CAAA42307-AK AA42307-K2 CAAA42307-K2
Gold plating solution ∼3.7 g/l metal content, electroless
Gold plating solution
Applications: For coating nickel or copper


Storage Temperature: Ambient
MDL Number: MFCD00003436
UN: 2922
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty