Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tricosachromium hexacarbide ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Chromium carbide

42313.3 42313-18 42313.09 42313-09 42313.18 42313-30
AA42313-18EA 91.59 USD InStock
CAAA42313-18 AA42313-18 AA42313-09 CAAA42313-09 CAAA42313-30 AA42313-30
Tricosachromium hexacarbide ≥99.5% (metals basis)
Tricosachromium hexacarbide


Formula: C₆Cr₂₃
Storage Temperature: Ambient
MDL Number: MFCD00678358
CAS Number: 12105-81-6
EINECS: 235-160-2
UN: 3178
ADR: 4.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty