Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Decamethylcyclopentasiloxane 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Silicone fluid DC 245 Volasil 245 Dow Corning 245® fluid Cyclopentasiloxane

42412-A1 042412-18 042412-30 042412-A1 42412-30 42412-18
AA42412-30EA 88.2 USD InStock
CAAA42412-30 AA42412-30 CAAA42412-A1 AA42412-A1 CAAA42412-18 AA42412-18
Decamethylcyclopentasiloxane 97%
Decamethylcyclopentasiloxane


Formula: C₁₀H₃₀O₅Si₅
MW: 370.77 g/mol
Boiling Pt: 210 °C
Melting Pt: –44 °C
Density: 0.959
Flash Pt: 76 °C (169 °F)
Storage Temperature: Ambient
MDL Number: MFCD00046966
CAS Number: 541-02-6
EINECS: 208-764-9
Merck Index: 12,02905
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty