Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Mercaptopropyl(dimethoxy)methylsilane ≥95%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 3-(Dimethoxymethylsilyl)-1-propanethiol3-Mercaptopropylmethyldimethoxysilane

42426-K2 42426-AK 042426.AB 042426.AK 42426-K2 42426-AD 42426-AB 042426.AD
AA42426-ABEA 25.09 USD InStock
CAAA42426-AB AA42426-AB CAAA42426-AD AA42426-AD CAAA42426-K2 AA42426-K2 CAAA42426-AK AA42426-AK
3-Mercaptopropyl(dimethoxy)methylsilane ≥95%
3-Mercaptopropyl(dimethoxy)methylsilane


Formula: C₆H₁₆O₂SSi
MW: 180.34 g/mol
Boiling Pt: 96 °C (30 mmHg)
Density: 1,000
Flash Pt: 93 °C (199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00053923
CAS Number: 31001-77-1
EINECS: 250-426-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty