Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), flakes -150 mesh, Thickness 1.2 µm Type 316-L

Supplier: Thermo Scientific Chemicals

042486-30 42486-30 042486-A1 42486-A1
AA42486-30EA 48.09 USD InStock
CAAA42486-30 AA42486-30 CAAA42486-A1 AA42486-A1
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), flakes -150 mesh, Thickness 1.2 µm Type 316-L
Stainless steel


Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty