Synonyms:
Zirconium dioxide
042493.A1
42493.3
42493-30
42493-A1
AA42493-30EA
84.79
USD
InStock
CAAA42493-30
CAAA42493-A1
AA42493-30
AA42493-A1
Zirconium(IV) oxide cement
Zirconium (IV) oxide
One component adhesive. Bonds to Zirconium oxide and Silicon. Exhibit good dielectric properties