Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000

Supplier: Thermo Scientific Chemicals

042502-AK 042502-K2 042502-AD 42502-AK 42502-AD 42502-K2
AA42502-ADEA 26.69 USD InStock
AA42502-AD CAAA42502-AD AA42502-AK CAAA42502-AK AA42502-K2 CAAA42502-K2
Polydimethylsiloxane, trimethylsiloxy terminated, M.W. 9000
Polydimethylsiloxane, trimethylsiloxy terminated
Viscosity 200 centistokes


Formula: (CH₃)₃SiO(Si(CH₃)₂O)nSi(CH₃)₃
Boiling Pt: >205 °C
Melting Pt: < –40 °C
Density: 0,968
Flash Pt: 315 °C (599 °F)
Storage Temperature: Ambient
MDL Number: MFCD00148360
CAS Number: 9016-00-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty