Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

42527-06 42527-01 42527-03 042527-03 42527-01 042527-06
AA42527-03EA 162.99 USD InStock
CAAA42527-06 AA42527-03 AA42527-06 CAAA42527-01 AA42527-01 CAAA42527-03
Dihydrogen hexabromoosmate(IV) hydrate ≥99.9% (metals basis)
Dihydrogen hexabromoosmate(IV) hydrate


Formula: H₂OsBr₆.xH₂O
MW: 352.02 g/mol (anhy)
Melting Pt: Dec.
Storage Temperature: Ambient
MDL Number: MFCD01863474
UN: 3260
ADR: 8,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty