042529-06
42529-03
042529-03
42529-06
AA42529-06EA
835.99
USD
InStock
AA42529-06
AA42529-03
CAAA42529-06
CAAA42529-03
Palladium (II) sulfate dihydrate ≥99% (metals basis)
Palladium (II) sulphate dihydrate
Soluble in cold water; decomposes in hot water