Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Apatite naturally occurring mineral, grains ∼ 0.06-0.19 in

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Calcium fluorophosphateApatiteFluorapatite

042535-06 042535-03 42535-03 42535-06 42535-14 042535-14
AA42535-03EA 34 USD InStock
AA42535-03 AA42535-14 CAAA42535-03 CAAA42535-14 AA42535-06 CAAA42535-06
Apatite naturally occurring mineral, grains ∼ 0.06-0.19 in
Apatite
Apatite is one of a few minerals produced and used by biological micro-environmental systems. The primary use of apatite is in the manufacture of fertilizer, it is a source of phosphorus. It is occasionally used as a gemstone. Green and blue varieties in finely divided form, are pigments with excellent covering power.

Sp. Gr. 3.1-3.2. Store away from strong oxidizing agents. Store in cool, dry conditions.


Formula: Ca₅FO₁₂P₃
MW: 504.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00285511
CAS Number: 1306-05-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty