Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Total Organic Carbon (TOC), Standard Solution, Specpure®, 1000µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
42562-AP 42562-AE 042562-AP 042562-AE
AA42562-APEA 398.8 USD InStock
AA42562-AP AA42562-AE CAAA42562-AE CAAA42562-AP
Total Organic Carbon (TOC), Standard Solution, Specpure®, 1000µg/ml, Thermo Scientific
Standards Environmental Analysis Standards
Standard for TOC based on potassium hydrogen phthalate (KHC₈H₄O₄).

Physical Form: Liquid
MDL No.: MFCD00133992

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty