Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Acetate, Ion chromatography standard solution, Specpure®, CH₃CO₂⁻ 1000 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
042563.AP 42563-AE 042563-AE 42563-AP
AA42563-AEEA 120.72 USD InStock
CAAA42563-AE AA42563-AE AAA42563-AP CAAAA42563-AP
Acetate, Ion chromatography standard solution, Specpure®, CH₃CO₂⁻ 1000 µg/ml, Thermo Scientific
Standards Anionic Standards Acetate Standards
Guaranteed stable for six months from the ship date

Molecular Formula: CH3CO2Na in H2O
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00282007

Keep Refrigerated

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty