Synonyms:
Polyaniline EB
, PAni EB
42581-03
42581.03
42581.06
42581-06
42581-14
42581.14
AA42581-06EA
203.95
USD
InStock
AA42581-06
CAAA42581-06
AA42581-03
CAAA42581-03
AAA42581-14
CAAAA42581-14
Polyaniline, emeraldine base
Polyaniline, emeraldine base
Applications: Conductive polymerPolyaniline is used in chemical vapor sensors, supercapacitors and biosensors. It is also useful for manufacture of electrical conducting yarns, antistatic coatings, electromagnetic shielding, flexible electrodes and printed circuit board. Further, it is used in antistatic and ESD coatings and corrosion protection.