Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dioctyldichlorosilane 97%

Supplier: Thermo Scientific Chemicals
Danger

42599-09 42599.09 42599.18 42599-18
AA42599-18EA 311.99 USD InStock
CAAA42599-18 AA42599-18 CAAA42599-09 AA42599-09
Dioctyldichlorosilane 97%
Dioctyldichlorosilane


Formula: C₁₆H₃₄Cl₂Si
Boiling Pt: 145 °C (0.2 mmHg)
Density: 0.940
Storage Temperature: Ambient
MDL Number: MFCD01074472
CAS Number: 18416-07-4
UN: 2987
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty