Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Octanesulfonic acid sodium salt, hydrate 99+% for HPLC

Supplier: Thermo Scientific Chemicals
Synonyms: Sodium 1-octansulphonate monohydrate

042636-06 42636-06 042636-22 42636-14 42636-22 042636-14
AA42636-06EA 50.19 USD InStock
AA42636-06 CAAA42636-06 CAAA42636-14 CAAA42636-22 AA42636-22 AA42636-14
1-Octanesulfonic acid sodium salt, hydrate 99+% for HPLC
1-Octanesulphonic acid sodium salt
Ion-pair reagent for HPLC analysis

Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent.


Formula: C₈H₁₇NaO₃S·1H₂O
MW: 216.28 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007544
CAS Number: 5324-84-5
EINECS: 226-195-4
Order Now
Specification Test Results
Loss on drying 6.9-8.5%
10% water solutionClear Pass Test.
colorless 0.4% EtOH/MeOHClear Pass Test
clear0.4% EtOH/MeOH Clear
IR identification Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty