Would you like to log in again?
Amberlite® IRA-958 (Cl), Amberlite™ ion exchange resin
Supplier: Thermo Scientific Chemicals
042702.A1 42702-30 042702.A7 42702-A1 42702.3 42702-A7
AA42702-A1EA 117.99 USD InStock
CAAA42702-A1 AA42702-A1 CAAA42702-A7 AA42702-A7 CAAA42702-30 AA42702-30
Amberlite® IRA-958 (Cl), Amberlite™ ion exchange resin
Amberlite® IRA-958 (Cl)
Uses: Removal of color, organics, and cyanide
Acrylic-DVB; Strongly basic anion exchanger with a quaternary ammonium functionality - chloride form
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty