Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 4.5 micron

Supplier: Thermo Scientific Chemicals

42715-AA 042715.AA
AA42715-AAEA 644.16 USD InStock
AA42715-AA CAAA42715-AA
Poly(styrene) 2,5% (w/w) dispersion in water, latex microsphere 4.5 micron
Poly(styrene)


Formula: (C₈H₈)n
MW: 125,000-250,000
Storage Temperature: Refrigerator
MDL Number: MFCD00084450
CAS Number: 9003-53-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty