Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrakis(dimethylamino)germanium(IV) ≥99% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

42760-ME 42760-04 42760-09 042760.ME 42760-04 42760.09
AA42760-MEEA 99.29 USD InStock
CAAA42760-ME CAAA42760-09 AA42760-ME AA42760-09 CAAA42760-04 AA42760-04
Tetrakis(dimethylamino)germanium(IV) ≥99% (metals basis)
Tetrakis(dimethylamino)germanium(IV)
Applications: Preparation of nitrides


Formula: C₈H₂₄GeN₄
MW: 248.90 g/mol
Boiling Pt: 203 °C
Melting Pt: 14 °C
Density: 1,07
Flash Pt: 34 °C (93 °F)
Storage Temperature: Ambient
MDL Number: MFCD01863476
CAS Number: 7344-40-3
UN: 3129
ADR: 4.3,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty