Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetraethylammonium hydroxide 35% (w/w) in aqueous solution, Reagent Grade

Supplier: Thermo Scientific Chemicals
Danger

043023-22 43023-22 043023-36 043023-14 43023-36 43023-14
AA43023-22EA 59.29 USD InStock
CAAA43023-22 AA43023-22 CAAA43023-36 CAAA43023-14 AA43023-36 AA43023-14
Tetraethylammonium hydroxide 35% (w/w) in aqueous solution, Reagent Grade
Tetraethylammonium hydroxide


Formula: (C₂H₅)₄N(OH)
MW: 147.26 g/mol
Boiling Pt: Dec.
Density: 1.023
Storage Temperature: Ambient
MDL Number: MFCD00009024
CAS Number: 77-98-5
EINECS: 201-073-3
UN: 3267
ADR: 8,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty