Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methyl-2-pentanone 99+% for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methyl Isobutyl KetoneIsobutyl Methyl KetoneMethyl iso-Butyl KetoneMIK

043170-K2 043170-M1 043170-K7 043170-M6
AA43170-M6EA 565 USD InStock
AA43170-M6 AA43170-K2 AA43170-K7 AA43170-M1
4-Methyl-2-pentanone 99+% for HPLC
4-Methyl-2-pentanone


Formula: (CH₃)₂CHCH₂COCH₃
MW: 100.16 g/mol
Boiling Pt: 117…118 °C
Melting Pt: –84 °C
Density: 0.801
Flash Pt: 13 °C (55 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008938
CAS Number: 108-10-1
EINECS: 203-550-1
UN: 1245
ADR: 3,II
Merck Index: 13,05227
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/40/739340.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty