Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethylaluminium ≥94%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Triethylaluminum

43243-22 43243-22
AA43243-22EA 398.99 USD InStock
AA43243-22 CAAA43243-22
Triethylaluminium ≥94%
Triethylaluminium
Catalyst for polymerization of olefins, intermediate
Pyrophoric. Packaged in non-returnable cylinders

Catalyst for polymerization of olefins, intermediate


Formula: C₆H₁₅Al
MW: 114.17 g/mol
Boiling Pt: 128…130 °C (50 mmHg)
Melting Pt: –50 °C
Density: 0.835
Flash Pt: 18 °C (–1 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009015
CAS Number: 97-93-8
EINECS: 202-619-3
UN: 3394
ADR: 4.2,I
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty