Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(±)-2-Butanol, anhydrous 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: sec-Butyl alcohol(±)-2-Hydroxy butane(±)-sec-Butyl alcohol(±)-sec-Butanolsec-Butanol(±)-Methyl ethyl carbinol(±)-Ethyl methyl carbinol

43315-K2 43315-AK 43315-K2 43315-AK
AA43315-AKEA 74.94 USD InStock
CAAA43315-K2 AA43315-AK AA43315-K2 CAAA43315-AK
(±)-2-Butanol, anhydrous 99%
(±)-2-Butanol


Formula: H₃CCH₂CH(OH)CH₃
MW: 74.12 g/mol
Boiling Pt: 98…101 °C
Melting Pt: –115 °C
Density: 0.808
Flash Pt: 26 °C (79 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004569
CAS Number: 78-92-2
EINECS: 201-158-5
UN: 1120
ADR: 3,II
Merck Index: 13,01540
https://us-legacy.vwr.com/stibo/web/std.lang.all/03/13/710313.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty