Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gold trichloride ≥99.99% (metals basis) Au ≥ 64.4%, Premion®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Gold(III) chlorideGold (III) chloride

43360-03 43360-03 43360-06 43360-06
AA43360-06EA 886.99 USD InStock
AA43360-06 CAAA43360-06 AA43360-03 CAAA43360-03
Gold trichloride ≥99.99% (metals basis) Au ≥ 64.4%, Premion®
Gold trichloride


Formula: AuCl₃
MW: 303.32 g/mol
Melting Pt: 180 °C (subl.)
Density: 3.90
Storage Temperature: Ambient
MDL Number: MFCD00014172
CAS Number: 13453-07-1
EINECS: 236-623-1
UN: 3260
ADR: 8,III
Merck Index: 13,04536
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty