Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Methyl levulinate 99+%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 4-Ketovaleric acid methyl ester 4-Oxopentanoic acid methyl ester 4-Oxovaleric acid methyl ester Levulinic acid methyl ester Methyl 4-oxovalerate

43397-36 43397-22 43397-14 43397-14 43397-22 43397-36
AA43397-14EA 55.69 USD InStock
CAAA43397-36 CAAA43397-14 AA43397-14 AA43397-36 CAAA43397-22 AA43397-22
Methyl levulinate 99+%
Methyl levulinate


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 191 °C
Melting Pt: –24 °C
Density: 1,05
Flash Pt: 72 °C (161.6 °F)
Storage Temperature: Ambient
MDL Number: MFCD00017499
CAS Number: 624-45-3
EINECS: 210-846-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty