Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydroxypropyl cellulose, M.W. 100000, 75 - 150 mPa.s (5 % in H₂O)

Supplier: Thermo Scientific Chemicals

43400-22 43400-36 43400-36 43400-22 43400-14 43400-A3 43400-A3 43400-14
AA43400-36EA 205.99 USD InStock
CAAA43400-22 AA43400-36 AA43400-14 AA43400-A3 CAAA43400-36 CAAA43400-14 AA43400-22 CAAA43400-A3
Hydroxypropyl cellulose, M.W. 100000, 75 - 150 mPa.s (5 % in H₂O)
Hydroxypropyl cellulose
Uses: Thickener, binder, and emulsifier
Average MW 100,000; viscosity (5 wt% aqueous solution, 25°) 75-150 cps; As ³3ppm, Pb ³10ppm
Soluble in polar organic solvents, in water at room temperature


Storage Temperature: Ambient
MDL Number: MFCD00132688
CAS Number: 9004-64-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty