Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Gallium trinitrate solution (9 - 10% Ga) (w/w)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Gallium(III) nitrateGallium (III) nitrate

43484-30 43484-30 43484-18 43484-18
AA43484-30EA 504.99 USD InStock
AA43484-30 CAAA43484-18 AA43484-18 CAAA43484-30
Gallium trinitrate solution (9 - 10% Ga) (w/w)
Gallium trinitrate
contains 25-30% HNO₃


Formula: GaN₃O₉
MW: 255.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011016
CAS Number: 13494-90-1
EINECS: 236-815-5
UN: 3218
ADR: 5.1,III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty