Would you like to log in again?
Fused Quartz Crucible and Lid, Thermo Scientific
Supplier: Thermo Scientific
045555-KG 043498-KG 45555-KG
AA45555-KGEA 109.99 USD InStock
CAAA45555-KG AA45555-KG AA43498-KG
Fused Quartz Crucible and Lid, Thermo Scientific
Crucibles
Fused quartz has many desirable properties including high chemical purity, high corrosion resistance, high melting point, extreme hardness, low coefficient of thermal expansion, excellent electrical insulation qualities, and optical transmission from ultra-violet to infra-red
Crucible and lid are manufactured by fusing naturally occurring crystalline silica.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty