Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polyaniline, emeraldine salt from p-Toluenesulfonic acid

Supplier: Thermo Scientific Chemicals
Synonyms: Polyaniline ESPAni ES

43683-18 43683-18 43683-09 43683-09 43683-30
AA43683-18EA 448.17 USD InStock
AA43683-18 CAAA43683-09 CAAA43683-18 AA43683-09 AAA43683-30
Polyaniline, emeraldine salt from p-Toluenesulfonic acid
Polyaniline, emeraldine salt
Applications: Polyaniline is used in chemical vapor sensors, supercapacitors and biosensors. It is also useful for manufacture of electrical conducting yarns, antistatic coatings, electromagnetic shielding, flexible electrodes and printed circuit board. Further, it is used in antistatic and ESD coatings and corrosion protection.


Formula: C₃H₅BrClN₃
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00284320
CAS Number: 25233-30-1
Order Now
Specification Test Results
Form Powder

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty