Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform ≥99.5% stabilized for HPLC

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

Stabilization: Stabilised with amylene

43685-K2 43685-K7 43685-K7 43685-M1 43685-M6 43685-M1 43685-M6 43685-K2
AA43685-K7EA 177.55 USD InStock
AA43685-K7 AA43685-K2 CAAA43685-M6 AA43685-M6 CAAA43685-K7 AA43685-M1 CAAA43685-K2 CAAA43685-M1
Chloroform ≥99.5% stabilized for HPLC
Chloroform


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 60.5…61.5 °C
Melting Pt: –63 °C
Density: 1.492
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://us-legacy.vwr.com/stibo/web/std.lang.all/69/36/726936.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty