Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Niobium(V) chloride ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Niobium pentachloride

43729-22 43729-22 43729-14 43729-14 43729-06 43729-06
AA43729-06EA 114.99 USD InStock
AA43729-06 CAAA43729-06 AA43729-14 AA43729-22 CAAA43729-14 CAAA43729-22
Niobium(V) chloride ≥99.999% (metals basis), Puratronic®
Niobium (V) chloride


Formula: Cl₅Nb
MW: 270.17 g/mol
Boiling Pt: 248.2 °C
Melting Pt: 208.3 °C
Density: 2.74
Storage Temperature: Ambient
MDL Number: MFCD00011127
CAS Number: 10026-12-7
EINECS: 233-059-8
UN: 3260
ADR: 8,III
Merck Index: 12,06649
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty