Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ninhydrin, REacton® ACS

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Indanetrione hydrate

43846-36 43846-22 43846-14 43846-36 43846-14 43846-22
AA43846-14EA 101.88 USD InStock
AA43846-14 AA43846-36 AA43846-22 CAAA43846-22 CAAA43846-36 CAAA43846-14
Ninhydrin, REacton® ACS
Ninhydrin
Reagent for determination of amino acids and amine containing compounds, as an intermediate.


Formula: C₉H₆O₄
MW: 178.14 g/mol
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 251 °C (dec.)
Density: 0.862 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003791
CAS Number: 485-47-2
EINECS: 207-618-1
Merck Index: 13,06582
https://us-legacy.vwr.com/stibo/web/std.lang.all/70/01/2297001.jpg
Order Now
Specification Test Results
Appearance White to brownish-white crystals
Identification and melting point Pass test.
Solubility Pass test
Sensitivity to amino acids Pass test

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty