Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Naphthyl sulfate potassium salt ≥98%

Supplier: Thermo Scientific Chemicals

44061-MD 44061-03 44061-MD 44061-03
AA44061-MDEA 49.79 USD InStock
AA44061-MD CAAA44061-MD AA44061-03 CAAA44061-03
2-Naphthyl sulfate potassium salt ≥98%
2-Naphthyl sulfate potassium salt


Formula: C₁₀H₇KO₄S
MW: 262.33 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004059
CAS Number: 1733-89-7
EINECS: 217-072-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty