Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)

Supplier: Thermo Scientific Chemicals
Synonyms: Nadide phosphate monosodium salt NADP-Na Nicotinamide-adenine dinucleotide phosphate monosodium salt NADP monosodium salt

44126-MD 44126-03 44126-03 44126-MD
AA44126-MDEA 126.99 USD InStock
AA44126-MD CAAA44126-MD AA44126-03 CAAA44126-03
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)
ß-Nicotinamide adenine dinucleotide phosphate (NADP-Na, oxidized form)


Formula: C₂₁H₂₇N₇NaO₁₇P₃
MW: 765.39 g/mol
Melting Pt: 175 to 178 °C
Storage Temperature: Freezer
MDL Number: MFCD00036973
CAS Number: 1184-16-3
EINECS: 214-664-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty