Synonyms:
Bis(1,5-cyclooctadiene)nickel(0) (Wax encapsulated) (ca. 0.05mmol/capsule)
, Ni(COD)2
44157-09
44157-04
44157-09
44157-04
AA44157-04EA
113.54
USD
InStock
CAAA44157-04
AA44157-04
CAAA44157-09
AA44157-09
Bis(1,5-cyclooctadiene)nickel(0) ≥96%
Bis(1,5-cyclooctadiene)nickel (0)
Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene
Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene