Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(1,5-cyclooctadiene)nickel(0) ≥96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bis(1,5-cyclooctadiene)nickel(0) (Wax encapsulated) (ca. 0.05mmol/capsule)Ni(COD)2

44157-09 44157-04 44157-09 44157-04
AA44157-04EA 113.54 USD InStock
CAAA44157-04 AA44157-04 CAAA44157-09 AA44157-09
Bis(1,5-cyclooctadiene)nickel(0) ≥96%
Bis(1,5-cyclooctadiene)nickel (0)
Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene

Catalyst for isomerization and hydrosilyltion of unsaturated compounds. Co-catalyst for oligomerization/cyclooligomerization or polymerization of alkenes, butadiene


Formula: C₁₆H₂₄Ni
MW: 275.06 g/mol
Melting Pt: 60 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00058902
CAS Number: 1295-35-8
EINECS: 215-072-0
UN: 1325
ADR: 4.1,II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty