Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Boron (≥96% ¹⁰B) (metals basis), crystalline powder -60+100 mesh

Supplier: Thermo Scientific Chemicals
Warning

44256-14 44256-06 44256-06 44256-14
AA44256-14EA 1570.99 USD InStock
AA44256-14 CAAA44256-06 CAAA44256-14 AA44256-06
Boron (≥96% ¹⁰B) (metals basis), crystalline powder -60+100 mesh
Boron
Neutron absorber


Formula: B
MW: 10.81 g/mol
Density: 1,73 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00143869
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1,III
Merck Index: 13,01333
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty