Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrapotassium diphosphate, anhydrous ≥96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Potassium diphosphate Tetrapotassium diphosphate tetra-Potassium pyrophosphate

44269-A1 44269-A7 44269-A7 44269-A1
AA44269-A7EA 273.26 USD InStock
CAAA44269-A7 AA44269-A7 AA44269-A1 CAAA44269-A1
Tetrapotassium diphosphate, anhydrous ≥96%
tetra-Potassium diphosphate


Formula: K₄O₇P₂
MW: 330.34 g/mol
Boiling Pt: Dec.
Melting Pt: 1090 °C
Density: 2.33
Storage Temperature: Ambient
MDL Number: MFCD00011393
CAS Number: 7320-34-5
EINECS: 230-785-7
Merck Index: 12,07833
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty