Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cobalt(II) acetate tetrahydrate ≥99.999% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Acetic acid cobalt (II) salt tetrahydrate Cobalt diacetate tetrahydrate Cobaltous acetate tetrahydrate

44345-04 44345-04 44345-09 44345-18 44345-18 44345-09
AA44345-09EA 268.99 USD InStock
AA44345-09 AA44345-18 CAAA44345-09 CAAA44345-04 CAAA44345-18 AA44345-04
Cobalt(II) acetate tetrahydrate ≥99.999% (metals basis)
Cobalt (II) acetate tetrahydrate


Formula: (H₃CCOO)₂Co·4H₂O
MW: 249.08 g/mol
Melting Pt: ∼140 °C (-4H₂O)
Density: 1.71
Storage Temperature: Ambient
MDL Number: MFCD00149569
CAS Number: 6147-53-1
EINECS: 200-755-8
UN: 3077
ADR: 9,III
Merck Index: 12,02494
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty