Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Hydroxy-5-methylisoxazole 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hymexazol

44450-06 44450-22 44450-06 44450-14 44450-22 44450-14
AA44450-06EA 53.69 USD InStock
AA44450-06 CA44450-06 AA44450-14 CAAA44450-22 AA44450-22 CAAA44450-14
3-Hydroxy-5-methylisoxazole 97%
3-Hydroxy-5-methylisoxazole


Formula: C₄H₅NO₂
MW: 99.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00144468
CAS Number: 10004-44-1
EINECS: 233-000-6
Order Now

 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty