Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

HSG-75, Electrically conductive heat sink grease, Ag ≈75%, Thermo Scientific

Supplier: Thermo Scientific
44590-MJ 44590-MJ 44590-O0 044590-O0
AA44590-O0EA 174.99 USD InStock
CAAA44590-O0 AA44590-O0 CAAA44590-MJ AA44590-MJ
HSG-75, Electrically conductive heat sink grease, Ag ≈75%, Thermo Scientific
Lubricants
Thermally and electrically conductive heat sink grease applied to the base and mounting stud of transistors, diodes and rectifiers, as a coupling agent to reduce thermal and electrical contact resistance

Physical Form: Viscous liquid
Appearance: Grey
UN No.:UN3082

Applications: A high loading of proprietary silver flake provides excellent electrical conductivity properties. This is ideal for mounting sputter targets where thermal or thermal plus electrical conductivity are needed.

Ideal for sputtering target applications

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty